Invitrogen
Manufacturer Code:PA5117266
Catalog # PIPA5117266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphorylated peptide around S1490 of human LRP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADCAD2 C030016K15Rik Cd crooked FLJ90062 FLJ90421 Gw LDL receptor related protein 6 low density lipoprotein receptor-related protein 6 low density lipoprotein receptor-related protein 6 low density lipoprotein-related protein 6 low density lipoprotein-related protein 6 low-density lipoprotein receptor-related protein 6 lpr6 Lrp6 LRP-6 ska<m26Jus> ska26 skax26 STHAG7; low density lipoprotein receptor-related protein 6; Low-density lipoprotein receptor-related protein 6; LRP-6
Gene Aliases: ADCAD2; LRP6; STHAG7
UniProt ID: (Human) O75581
Entrez Gene ID: (Human) 4040
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.