Invitrogen
Manufacturer Code:PA538664
Catalog # PIPA538664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human LEF-1 around the phosphorylation site of Ser42 (E-I-SP-H-P) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LEF-1; LEF1 lymphoid enhancer-binding factor 1 HGNC:6551 DKFZp586H0919 TCF1ALPHA lymphoid enhancer binding factor-1; lymphoid enhancer binding factor 1 short isoform; Lymphoid enhancer-binding factor 1; T cell-specific transcription factor 1-alpha; TCF1-alpha
Gene Aliases: 3000002B05; AI451430; LEF-1; LEF1; TCF10; TCF1ALPHA; TCF7L3
UniProt ID: (Human) Q9UJU2, (Rat) Q9QXN1, (Mouse) P27782
Entrez Gene ID: (Human) 51176, (Rat) 161452, (Mouse) 16842
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.