Invitrogen
Manufacturer Code:PA5105445
Catalog # PIPA5105445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human LDHA around the phosphorylation site of Tyr10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 19 protein; cell proliferation-inducing gene 19 protein epididymis secretory sperm binding protein Li 133P GSD11 HEL-S-133P l7R2 Lactate Dehydrogenase lactate dehydrogenase 1 A chain lactate dehydrogenase A lactate dehydrogenase A4 lactate dehydrogenase A-like lactate dehydrogenase M lactate dehydrogenase-A lactate dehydrogenase-M Lactic Dehydrogenase LDH LDH muscle subunit Ldh1 Ldh-1 Ldha LDH-A ldha protein ldha1 LDH-A4 muscle-specific isozyme glycolytic enzyme LDHM LDH-M L-lactate dehydrogenase L-lactate dehydrogenase A chain M-LDH OTTHUMP00000165206 PIG19 proliferation-inducing gene 19 renal carcinoma antigen NY-REN-59; epididymis secretory sperm binding protein Li 133P; L-lactate dehydrogenase A chain; L-lactate dehydrogenase C chain; lactate dehydrogenase 1, A chain; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase A-like; lactate dehydrogenase A4; lactate dehydrogenase C variant 3; lactate dehydrogenase M; lactate dehydrogenase-A; LDH muscle subunit; LDH testis subunit; LDH-A; LDH-C; LDH-M; LDH-X; M-LDH; proliferation-inducing gene 19; Renal carcinoma antigen NY-REN-59
Gene Aliases: GSD11; HEL-S-133P; l7R2; Ldh-1; Ldh1; Ldh3; LDHA; LDHM; PIG19
UniProt ID: (Human) P00338, (Rat) P04642, (Mouse) P06151
Entrez Gene ID: (Human) 3939, (Rat) 24533, (Mouse) 16828, (Rat) 29634
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.