Invitrogen
Manufacturer Code:PA5105148
Catalog # PIPA5105148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Ku70 around the phosphorylation site of Ser520. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-deoxyribose-5-phosphate lyase Ku70; 5'-deoxyribose-5-phosphate lyase Ku70 5'-dRP lyase Ku70 5'-dRP/AP lyase Ku70 70 kDa subunit of Ku antigen 70kDa ATP-dependent DNA helicase 2 subunit 1 ATP-dependent DNA helicase II 70 kDa subunit ATP-dependent DNA helicase II 70 kDa subunit CT CTA-216E10.7 CTC box binding factor 75 kDa subunit CTC box-binding factor 75 kDa subunit CTC75 CTCBF DNA repair protein XRCC6 G22p1 Ku autoantigen p70 subunit ku autoantigen protein p70 homolog Ku autoantigen 70kDa Ku p70 KU70 Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa) Kup70 Lupus Ku autoantigen protein p70 ML8 OTTHUMP00000028581 thyroid autoantigen thyroid autoantigen 70 kDa thyroid autoantigen 70kD (Ku antigen) thyroid autoantigen 70kDa (Ku antigen) Thyroid-lupus autoantigen thyroid-lupus autoantigen p70 TLAA X-ray repair complementing defective repair in Chinese hamster cells 6 X-ray repair cross complementing 6 X-ray repair cross-complementing protein 6 XRCC6; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTCBF; DNA repair protein XRCC6; Ku autoantigen p70 subunit; Ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Lupus Ku autoantigen protein p70; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair cross-complementing protein 6
Gene Aliases: 70kDa; CTC75; CTCBF; G22P1; KU70; Kup70; ML8; TLAA; XRCC6
UniProt ID: (Human) P12956, (Mouse) P23475
Entrez Gene ID: (Human) 2547, (Rat) 25019, (Mouse) 14375
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.