Invitrogen
Manufacturer Code:MA528079
Catalog # PIMA528079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | KSR1S392-3A4 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Ser392 of human phospho KSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Kinase suppressor of Ras 1; mKSR1; Protein Hb
Gene Aliases: AW492498; B-KSR1; D11Bhm183e; D11Bhm184e; hb; KSR; KSR1; RSU2
UniProt ID: (Human) Q8IVT5, (Mouse) Q61097
Entrez Gene ID: (Human) 8844, (Mouse) 16706
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.