Invitrogen
Manufacturer Code:MA537446
Catalog # PIMA537446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JAK2Y10071008-PB6 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr1007/Tyr1008 of human phospho Jak2. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10.2 Fd17 JAK 2 Jak2 JAK-2 Janus kinase 2 Janus kinase 2 (a protein tyrosine kinase) JTK 10 JTK10 kinase Jak2 THCYT3 Tyrosine-protein kinase JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Tyrosine-protein kinase JAK2
Gene Aliases: JAK2; JTK10; THCYT3
UniProt ID: (Human) O60674
Entrez Gene ID: (Human) 3717
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.