Invitrogen
Manufacturer Code:MA536892
Catalog # PIMA536892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | Jak1Y10221023-F11 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr1022/1023 of human phospho-Jak1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AA960307 BAP004 C130039L05Rik EC 2.7.10.2 Jak1 JAK-1 JAK1A JAK1B Janus kinase 1 Janus protein tyrosine kinase 1 JTK3 kinase Jak1 Tyrosine-protein kinase JAK1; JAK-1; Janus kinase 1; Tyrosine-protein kinase JAK1
Gene Aliases: JAK1; JAK1A; JAK1B; JTK3
UniProt ID: (Human) P23458
Entrez Gene ID: (Human) 3716
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.