Invitrogen
Manufacturer Code:PA19593
Catalog # 01-674-467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Phosphorylated peptide (pT183-P-pY185) based on human JNK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun kinase 2; C-jun kinase JNK 55 MAPK Mitogen activated protein kinase SAPK PRKM Protein Kinase Mitogen Activated SAPK 1 Stress activated protein kinase J Phospho-JNK; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK 9; Mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; Mitogen-activated protein kinase 9; p54 gamma; p54-alpha; SAPK gamma; SAPK-alpha; SAPK1a; SAPK1c; stress activated protein kinase alpha II; stress-activated protein kinase 1; Stress-activated protein kinase 1a; Stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2
Gene Aliases: JNK; JNK-46; JNK-55; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK8; MAPK9; p54a; p54aSAPK; PRKM8; PRKM9; SAPK; SAPK1; SAPK1A; SAPK1C
UniProt ID: (Human) Q15713, (Human) P45984, (Rat) P49185, (Rat) P49186
Entrez Gene ID: (Human) 5599, (Human) 5601, (Rat) 116554, (Rat) 50658
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.