Invitrogen
Manufacturer Code:PA5117229
Catalog # PIPA5117229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding Y1007 of human JAK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10.2 Fd17 JAK 2 Jak2 JAK-2 Janus kinase 2 Janus kinase 2 (a protein tyrosine kinase) JTK 10 JTK10 kinase Jak2 THCYT3 Tyrosine-protein kinase JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Tyrosine-protein kinase JAK2
Gene Aliases: Fd17; JAK2; JTK10; THCYT3
UniProt ID: (Human) O60674, (Mouse) Q62120
Entrez Gene ID: (Human) 3717, (Mouse) 16452
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.