Invitrogen
Manufacturer Code:PA567508
Catalog # PIPA567508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human JAK2/JAK3 around the phosphorylation site of Tyr966/939. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JAK-2; JAK-3; JAK3B Breast-JAK3; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L-JAK; Leukocyte janus kinase; Tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3
Gene Aliases: fae; Fd17; JAK-3; JAK2; JAK3; JAK3_HUMAN; JAKL; JTK10; L-JAK; LJAK; RATJAK3; THCYT3; wil
UniProt ID: (Human) O60674, (Human) P52333, (Rat) Q62689, (Mouse) Q62120, (Mouse) Q62137, (Rat) Q63272
Entrez Gene ID: (Human) 3717, (Human) 3718, (Rat) 24514, (Mouse) 16452, (Mouse) 16453, (Rat) 25326
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.