Invitrogen
Manufacturer Code:PA5117222
Catalog # PIPA5117222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding s724 of human IRE1alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9030414B18Rik AI225830 C85377 endoplasmic reticulum (ER) to nucleus signalling 1 endoplasmic reticulum to nucleus signaling 1 endoplasmic reticulum to nucleus signalling 1 endoplasmic reticulum-to-nucleus signaling 1 Endoribonuclease ER to nucleus signalling 1 ERN1 FLJ30999 hIRE1p inositol-requiring 1 inositol-requiring 1 alpha inositol-requiring enzyme 1 Inositol-requiring protein 1 IRE1 IRE1a IRE1a antibody Ire1alpha Ire1-alpha IRE1P IRE1p antibody LOC100357346 LOC417414 MGC163277 MGC163279 protein kinase/endoribonuclease RGD1559716 Serine/threonine-protein kinase Serine/threonine-protein kinase/endoribonuclease IRE1; endoplasmic reticulum (ER) to nucleus signalling 1; Endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; Inositol-requiring protein 1; Ire1-alpha; IRE1a; protein kinase/endoribonuclease; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1
Gene Aliases: 9030414B18Rik; AI225830; C85377; ERN1; hIRE1p; IRE1; IRE1a; Ire1alpha; IRE1P; RGD1559716
UniProt ID: (Human) O75460, (Mouse) Q9EQY0
Entrez Gene ID: (Human) 2081, (Mouse) 78943, (Rat) 498013
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.