Invitrogen
Manufacturer Code:700393
Catalog # PI700393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 97H9L7 |
Immunogen | A peptide corresponding to amino acids 1186-1197 and 1162-1173 of P06213 and P08069 respectively |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD220; CD221; IGF-I receptor; IGF1 Receptor IGF-I Receptor IGF1R CD221 IGFIR IGFR JTK13 IGF-I receptor insulin-like growth factor I receptor soluble IGF1R variant 1 soluble IGF1R variant 2 CD220 HHF5 INSR Insulin receptor; Insulin receptor; Insulin receptor subunit alpha; Insulin receptor subunit beta; Insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; IR; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Aliases: CD220; CD221; HHF5; IGF1R; IGFIR; IGFR; INSR; JTK13
UniProt ID: (Human) P08069, (Human) P06213
Entrez Gene ID: (Human) 3480, (Human) 3643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.