Invitrogen
Manufacturer Code:PA5105140
Catalog # PIPA5105140
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IGF-I Receptor around the phosphorylation site of Tyr1131. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4932439J01Rik A330103N21Rik CD220 CD221 D630014A15Rik D930020L01 HHF5 hyft IGF1 Receptor IGF-1 receptor IGF1 Receptor beta Igf1r IGF-1R IGF1R beta IGF-I receptor IGF-I Receptor beta IGFIR IGFIRC IGFR IGFR1 Insr insulin like growth factor 1 receptor insulin receptor insulin receptor precursor insulin receptor preproprotein Insulin receptor subunit alpha Insulin receptor subunit beta Insulin-like growth factor 1 receptor Insulin-like growth factor 1 receptor alpha chain Insulin-like growth factor 1 receptor beta chain insulin-like growth factor 1 receptor beta subunit insulin-like growth factor I receptor insulin-like growth factor receptor 1 insulin-like growth factor receptor type 1 insulin-like growth factor-I receptor insulin-like growth factor-I receptor precursor IR IR-A IR-B JTK13 line 186 soluble IGF1R variant 1 soluble IGF1R variant 2; CD220; CD221; IGF-1 receptor; IGF-I receptor; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; Insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; Insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; IR; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Aliases: 4932439J01Rik; A330103N21Rik; CD220; CD221; D630014A15Rik; D930020L01; HHF5; hyft; IGF-1R; IGF1R; IGFIR; IGFIRC; IGFR; Igfr1; INSR; IR; IR-A; IR-B; JTK13
UniProt ID: (Human) P08069, (Human) P06213, (Rat) P24062, (Mouse) Q60751, (Mouse) P15208
Entrez Gene ID: (Pig) 397350, (Human) 3480, (Human) 3643, (Pig) 396755, (Rat) 25718, (Mouse) 16001, (Rat) 24954, (Mouse) 16337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.