Invitrogen
Manufacturer Code:PA5105135
Catalog # PIPA5105135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Insulin Receptor around the phosphorylation site of Tyr1355. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4932439J01Rik CD 220 CD220 CD220 antigen CD220 beta CD221 D630014A15Rik HHF 5 HHF5 IGF-I receptor INSR insulin receptor insulin receptor preproprotein Insulin receptor subunit alpha Insulin receptor subunit beta IR IR 1 IR beta IR1 IR-1 IR-A IR-B; CD220; Insulin receptor; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR
Gene Aliases: CD220; HHF5; INSR
UniProt ID: (Human) P06213
Entrez Gene ID: (Human) 3643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.