Invitrogen
Manufacturer Code:710101
Catalog # PI710101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 8HCLC |
Immunogen | Phosphopeptide corresponding to amino acids 11531162 of human Insulin receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: CD220; CD220 HHF5 insulin receptor INSR CD221 IGF-I receptor insulin-like growth factor I receptor soluble IGF1R variant 1 soluble IGF1R variant 2; Insulin receptor; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR
Gene Aliases: CD220; HHF5; INSR
UniProt ID: (Human) P06213
Entrez Gene ID: (Human) 3643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.