Invitrogen
Manufacturer Code:MA514857
Catalog # PIMA514857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.10.3 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer177 of IKKß |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; I kappa B kinase 1; i kappa-B kinase alpha; I-kappa-B kinase 1; I-kappa-B kinase 2; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; I-kappa-B-kinase beta; IkappaB kinase alpha; IkB kinase alpha subunit; ikBKA; IKK alpha; IKK-A; IKK-a kinase; IKK-alpha; IKK-B; IKK-beta; IKK1; IKK2; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; Inhibitor of nuclear factor kappa-B kinase subunit alpha; Inhibitor of nuclear factor kappa-B kinase subunit alpha I-kappa-B kinase alpha IKK-A IKK-alpha IkBKA IkappaB kinase Conserved helix-loop-helix ubiquitous kinase I-kappa-B kinase 1 IKK1 Nuclear factor NF-kappa-B inhibitor kinase alpha NFKBIKA Transcription factor 16 TCF-16 CHUK IKKA TCF16; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKA; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase beta; Nuclear factor NFkappaB inhibitor kinase alpha; Serine/threonine protein kinase IKBKB; TCF-16; Transcription factor 16
Gene Aliases: AI132552; AI256658; AIM-1; CHUK; Chuk1; Fbx24; Fbxo24; IKBKA; IKBKB; IKK-2; IKK-alpha; IKK-beta; IKK1; IKK2; IKK[b]; IKKA; IKKB; IKKbeta; IMD15; NFKBIKA; NFKBIKB; TCF16
UniProt ID: (Bovine) Q95KV1, (Human) O15111, (Bovine) Q95KV0, (Human) O14920, (Mouse) O88351, (Rat) Q9QY78
Entrez Gene ID: (Bovine) 281073, (Human) 1147, (Mouse) 12675, (Rat) 309361, (Bovine) 281854, (Human) 3551, (Mouse) 16150, (Rat) 84351
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.