Invitrogen
Manufacturer Code:PA5104697
Catalog # PIPA5104697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IKK alpha/IKK beta around the phosphorylation site of Ser176/Ser177. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI132552 AI256658 AIM-1 CHUK Chuk1 component of inhibitor of nuclear factor kappa B kinase complex conserved helix-loop-helix ubiquitous kinase EC 2.7.11.10 Fbx24 Fbxo24 I kappa B kinase 1 i kappa-B kinase alpha IkappaB kinase I-kappa-B kinase 1 I-kappa-B kinase 2 IkappaB kinase alpha I-kappa-B kinase alpha I-kappa-B kinase-alpha I-kappa-B-kinase beta IkB kinase alpha subunit IkB kinase-alpha IkB kinase-beta ikBKA Ikbkb IKK alpha IKK[b] IKK1 IKK2 IKK-2 Ikka IKK-A IKK-a kinase IKK-alpha IKKB IKK-B IKKbeta IKK-beta IMD15 inhibitor of kappa light polypeptide gene enhancer in B-cells kinase beta inhibitor of kappaB kinase beta inhibitor of nuclear factor kappa B kinase beta subunit inhibitor of nuclear factor kappa B kinase subunit beta inhibitor of nuclear factor kappa-B kinase subunit alpha Inhibitor of nuclear factor kappa-B kinase subunit beta NFKBIKA NFKBIKB Nuclear factor NFkappaB inhibitor kinase alpha nuclear factor NF-kappa-B inhibitor kinase alpha Nuclear factor NF-kappa-B inhibitor kinase beta TCF16 TCF-16 TCF17 Transcription factor 16; Conserved helix-loop-helix ubiquitous kinase; I kappa B kinase 1; I-kappa-B kinase 1; I-kappa-B kinase 2; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; I-kappa-B-kinase beta; IkappaB kinase alpha; IkB kinase alpha subunit; IKK alpha; IKK-A; IKK-a kinase; IKK-B; IKK-beta; IKK1; IKK2; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; Inhibitor of nuclear factor kappa-B kinase subunit alpha; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKA; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase beta; Nuclear factor NFkappaB inhibitor kinase alpha; Serine/threonine protein kinase IKBKB; TCF-16; Transcription factor 16
Gene Aliases: AI132552; AI256658; AIM-1; CHUK; Chuk1; Fbx24; Fbxo24; IKBKA; IKBKB; IKK-2; IKK-alpha; IKK-beta; IKK1; IKK2; IKK[b]; IKKA; IKKB; IKKbeta; IMD15; NFKBIKA; NFKBIKB; TCF16
UniProt ID: (Human) O15111, (Human) O14920, (Mouse) O88351, (Rat) Q9QY78
Entrez Gene ID: (Human) 1147, (Mouse) 12675, (Rat) 309361, (Human) 3551, (Mouse) 16150, (Rat) 84351
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.