Invitrogen
Manufacturer Code:PA1087
Catalog # PIPA1087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide conjugated to KLH corresponding to residues GGRSRSG(pS)IVELI (414-426) of Human HTT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HD protein; HD protein homolog; Htt Huntingtin HD IT15; Huntingtin; Huntingtin, myristoylated N-terminal fragment; Huntington disease gene homolog; Huntington disease protein; huntington disease protein homolog
Gene Aliases: AI256365; C430023I11Rik; HD; Hdh; HTT; IT15
UniProt ID: (Human) Q9UQB7
Entrez Gene ID: (Human) 3064, (Mouse) 15194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.