Invitrogen
Manufacturer Code:PA517318
Catalog # PIPA517318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the amino terminus of histone H3 phosphorylated on Ser28 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG5825-PA; CG5825-PC; CG5825-PD; family 3B (H3.3B); H3 histone family, member A; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3A/B; H3.A; H3.B; H3L-like histone; His3.3A-PA; His3.3A-PC; His3.3A-PD; histone ; histone 1, H2ai; histone 1, H3a; histone cluster 1, H2ai; histone cluster 1, H2ai-like; histone cluster 1, H3a; histone cluster 3, H3; histone H3; Histone H3 class I; Histone H3 class II; Histone H3 containing protein; Histone H3 Histone 1 H3a Histone H3.1 histone H3/a HIST1H3A; Histone H3.1; Histone H3.2; histone H3.2-like; Histone H3.3; histone H3.3 ; Histone H3.3A; Histone H3.3B; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l; lamprey; lpy; unnamed protein product
Gene Aliases: 9430068D06Rik; CG5825; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; fb58e10; h3; H3 histone; H3-3A; H3-3B; h3-5; H3-I; H3-II; H3-III; H3-IV; H3-IX; H3-V; H3-VI; H3-VII; H3-VIII; H3-X; H3.1-221; H3.1-291; H3.1-I; h3.2a; H3.3; H3.3A; H3.3B; H3/A; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3F3; H3F3A; H3F3B; h3f3b.1; h3f3c; h3f3d; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3g; H3h; H3i; h3r; His-3.3A; His3.3; His3.3A; His3.3B; Hist1h2ai; Hist1h2ail; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; hist2h3; hist2h3c; I79_014844; PH3; PP781; wu:fb58e10; XELAEV_18028537mg; XELAEV_18044072mg; zgc:110292; zgc:56193; zgc:56418; zgc:64222; zgc:86731
UniProt ID: (Bovine) Q5E9F8, (Bovine) A4IFS1, (Human) P84243, (Chicken) P84247, (Mouse) P84244, (Rat) P84245, (Chicken) P84229, (Human) P68431, (Fruit fly) C0HL66, (Rat) Q6LED0, (Mouse) P68433, (Zebrafish) Q6PI20, (Xenopus) Q6PI79
Entrez Gene ID: (Bovine) 326601, (Bovine) 617051, (Human) 3020, (Chicken) 396233, (Human) 3021, (Mouse) 15078, (Chinese hamster) 100753137, (Mouse) 15081, (Rat) 117056, (Chicken) 768333, (Human) 8350, (Fruit fly) 33736, (Rat) 291159, (Mouse) 360198, (Rat) 679994, (Zebrafish) 406269, (Xenopus) 379757, (Xenopus) 399088
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.