Invitrogen
Manufacturer Code:PA517869
Catalog # PIPA517869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer10 of human histone H3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG5825-PA; CG5825-PC; CG5825-PD; H3 histone family, member A; H3.A; H3.B; H3L-like histone; His3.3A-PA; His3.3A-PC; His3.3A-PD; histone ; histone 1, H2ai; histone 1, H3a; histone cluster 1, H2ai; histone cluster 1, H2ai-like; histone cluster 1, H3a; histone cluster 3, H3; Histone H3; Histone H3 Histone 1 H3a Histone H3.1 histone H3/a HIST1H3A H3s10; Histone H3.1; histone H3.3; histone H3.3 ; Histone H3.3A; Histone H3.3B; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l
Gene Aliases: BUR5; CG5825; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; h3; H3 histone; H3.1-221; H3.1-291; H3.1-I; h3.2a; H3.3; H3.3A; H3/A; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3g; H3h; H3i; h3r; HHT1; HHT2; His-3.3A; His3.3; His3.3A; His3.3B; Hist1h2ai; Hist1h2ail; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; hist2h3; hist2h3c; N2749; PH3; SIN2; XELAEV_18028537mg; YBR010W; YBR0201; YNL031C
UniProt ID: (Baker's yeast) P61830, (Human) P68431, (Fruit fly) C0HL66, (Rat) Q6LED0, (Mouse) P68433
Entrez Gene ID: (Baker's yeast) 852295, (Human) 8350, (Fruit fly) 33736, (Rat) 291159, (Rat) 679994, (Mouse) 360198, (Xenopus) 399088
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.