Invitrogen
Manufacturer Code:PA538087
Catalog # PIPA538087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human HSL around the phosphorylation site of Ser855/554 (S-V-SP-E-A) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.79 Hormone sensitive lipase LIPE LIPS; hormone - sensitive lipase testicular isoform; Hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; lipase E; lipase, hormone-sensitive; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: 4933403G17Rik; AOMS4; FPLD6; HSL; LHS; LIPE
UniProt ID: (Human) Q05469, (Rat) P15304, (Mouse) P54310
Entrez Gene ID: (Human) 3991, (Rat) 25330, (Mouse) 16890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.