Invitrogen
Manufacturer Code:PA537597
Catalog # PIPA537597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of Serine 398(S-H-S(p)-R-Q) derived from Human Histamine H1 Receptor . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bordetella pertussis induced histamine sensitization; H1R; Histamine H1 receptor; histamine receptor, subclass H1; HRH1 H1 histamine receptor H1 receptor Histamine 1 receptor Histamine H1 receptor H1-R H1R Histamine H(1) receptor Histamine receptor H1 HisH1
Gene Aliases: Bphs; H1-R; H1R; HH1R; Hir; hisH1; HRH1
UniProt ID: (Human) P35367, (Mouse) P70174
Entrez Gene ID: (Human) 3269, (Mouse) 15465
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.