Invitrogen
Manufacturer Code:PA5105350
Catalog # PIPA5105350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HES1 around the phosphorylation site of Ser37. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHb39; BHLHB39 class B basic helix-loop-helix protein 39 FLJ20408 Hairy and enhancer of split 1 hairy and enhancer of split 1 (Drosophila) hairy homolog Hairy-like Hairy-like protein hes family bHLH transcription factor 1 Hes1 Hes-1 hHL HL HRY RHL transcription factor HES-1; Class B basic helix-loop-helix protein 39; Hairy and enhancer of split 1; Hairy homolog; Hairy-like protein; hHL; RHL; Transcription factor HES-1
Gene Aliases: BHLHB39; HES-1; HES1; HHL; HL; HRY
UniProt ID: (Human) Q14469, (Mouse) P35428, (Rat) Q04666
Entrez Gene ID: (Human) 3280, (Mouse) 15205, (Rat) 29577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.