Invitrogen
Manufacturer Code:PA538116
Catalog # PIPA538116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human HDAC5 around the phosphorylation site of Ser259 (T-A-SP-E-P) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-9; HD5; histone deacetylase 4; Histone deacetylase 5; Histone deacetylase 5 Short name&equalsHD5 Antigen NY-CO-9 HDAC-5 KIAA0600; histone deacetylase mHDA1
Gene Aliases: AI426555; HD5; Hdac4; HDAC5; KIAA0600; mHDA1; mKIAA0600; NY-CO-9
UniProt ID: (Human) Q9UQL6
Entrez Gene ID: (Human) 10014, (Mouse) 15184
Molecular Function: deacetylase hydrolase nucleic acid binding oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.