Invitrogen
Manufacturer Code:PA5105316
Catalog # PIPA5105316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HCN1 around the phosphorylation site of Ser846. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCNG-1; BCNG1 BCNG-1 brain cyclic nucleotide-gated channel 1 C630013B14Rik EIEE24 Hac2 HAC-2 Hcn1 hyperpolarization activated cyclic nucleotide gated potassium channel 1 hyperpolarization-activated cation channel 2 hyperpolarization-activated cyclic nucleotide-gated potassium channel 1 hyperpolarization-activated cyclic nucleotide-gated K+ 1 potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1; Brain cyclic nucleotide-gated channel 1; HAC-2; Hyperpolarization-activated cation channel 2; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Aliases: BCNG-1; BCNG1; C630013B14Rik; EIEE24; HAC-2; HAC2; HCN1
UniProt ID: (Human) O60741, (Mouse) O88704, (Rat) Q9JKB0
Entrez Gene ID: (Human) 348980, (Mouse) 15165, (Rat) 84390
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.