Invitrogen
Manufacturer Code:PA538497
Catalog # PIPA538497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human HCK around the phosphorylation site of Tyr410 (N-E-YP-T-A) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell/myeloid kinase; B-cell/myeloid kinase; BMK; Hematopoietic cell kinase; Hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9 p59Hck p61Hck hematopoietic cell kinase p59-HCK/p60-HCK tyrosine-protein kinase HCK; p56-HCK; p56-HCK/p59-HCK; p56Hck; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: AI849071; Bmk; HCK; Hctk; JTK9; p56Hck; p59Hck; p61Hck
UniProt ID: (Human) P08631, (Rat) P50545, (Mouse) P08103
Entrez Gene ID: (Human) 3055, (Rat) 25734, (Mouse) 15162
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.