Invitrogen
Manufacturer Code:PA538495
Catalog # PIPA538495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human GluR4 around the phosphorylation site of Ser862 (R-L-SP-I-T) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 4; GluA4; GluR-4; GluR-D; Glutamate receptor 4; Glutamate receptor 4 GluR-4 GluR4 AMPA-selective glutamate receptor 4 GluR-D Glutamate receptor ionotropic AMPA 4 GluA4 GRIA4 GLUR4; glutamate receptor channel alpha 4; Glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; spike wave 1
Gene Aliases: GluA4; Glur-4; GluR-D; GLUR4; GLUR4C; Gluralpha4; GLURD; GRIA4; spkw1
UniProt ID: (Human) P48058, (Mouse) Q9Z2W8
Entrez Gene ID: (Human) 2893, (Mouse) 14802
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.