Invitrogen
Manufacturer Code:PA5110124
Catalog # PIPA5110124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A phospho specific peptide corresponding to residues surrounding S845 of human GRIA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900051M01Rik AI853806 AMPA-selective glutamate receptor 1 Glr1 Glr-1 GluA1 GLUH 1 GLUH1 GluR K1 GLUR1 GluR-1 GluRA GluR-A GluRK1 GluR-K1 Glutamate receptor 1 Glutamate receptor ionotropic AMPA 1 GPRC1A Gria 1 GRIA1 GRM1A HBGR1 HGBR1 HIPA HIPA1 MGC13325 MGC133252 mGlu1 MGLUR1 MGLUR1A NMDA1 NMDAR1 NR1 OTTHUMP00000160643 OTTHUMP00000165781 OTTHUMP00000224241 OTTHUMP00000224242 OTTHUMP00000224243 RP11-350O14.1; AMPA 1; AMPA-selective glutamate receptor 1; GluA1; GluR-1; GluR-A; GluR-K1; Glutamate receptor 1; Glutamate receptor ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA 1
Gene Aliases: GluA1; GLUH1; GLUR1; GLURA; GRIA1; HBGR1
UniProt ID: (Human) P42261
Entrez Gene ID: (Human) 2890
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.