Invitrogen
Manufacturer Code:PA5105017
Catalog # PIPA5105017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FRS2 around the phosphorylation site of Tyr436. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4732458E18 C330018A15Rik FGFR signalling adaptor FGFR substrate 2 FGFR-signaling adaptor SNT Fibroblast growth factor receptor substrate 2 FRS1A FRS2 Frs2a Frs2alpha FRS2-alpha SNT SNT1 SNT-1 SNT2 Suc1-associated neurotrophic factor target 1; FGFR signalling adaptor; FGFR substrate 2; FGFR-signaling adaptor SNT; Fibroblast growth factor receptor substrate 2; FRS2-alpha; SNT-1; Suc1-associated neurotrophic factor target 1
Gene Aliases: 4732458E18; C330018A15Rik; FRS1A; FRS2; FRS2A; FRS2alpha; SNT; SNT-1; SNT1
UniProt ID: (Human) Q8WU20, (Mouse) Q8C180
Entrez Gene ID: (Human) 10818, (Mouse) 327826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.