Invitrogen
Manufacturer Code:711281
Catalog # PI711281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 12HCLC |
Immunogen | Peptide corresponding to Human FOXO4 (aa 193-201) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: AFX AFX1 MLLT7; Fork head domain transcription factor AFX1; Forkhead box protein O4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7
Gene Aliases: AFX; AFX1; FOXO4; MLLT7
UniProt ID: (Human) P98177
Entrez Gene ID: (Human) 4303
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.