Invitrogen
Manufacturer Code:PA537648
Catalog # PIPA537648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 197 (A-A-S(p)-M-D) derived from Human AFX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFX AFX1 Afxh Forkhead box protein O4 MLLT7 Putative forkhead domain AFX1 putative fork head domain AFX1; Afxh; Fork head domain transcription factor AFX1; Forkhead box protein O4; forkhead protein; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7
Gene Aliases: AFX; AFX1; Afxh; Fkhr3; FOXO4; MLLT7; RGD1561201
UniProt ID: (Human) P98177, (Mouse) Q9WVH3
Entrez Gene ID: (Human) 4303, (Rat) 302415, (Mouse) 54601
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.