Invitrogen
Manufacturer Code:PA599139
Catalog # PIPA599139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FER around the phosphorylation site of Tyr402. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV082135 C330004K01Rik c-FER EC 2.7.10.2 Feline encephalitis virus-related kinase FER FER fer (fms/fps related) protein kinase fer (fms/fps related) protein kinase testis specific 2 fer (fps/fes related) tyrosine kinase FER tyrosine kinase Fert Fert2 Flk Flk_retired Fujinami poultry sarcoma/Feline sarcoma-related protein Fer p47 p94-Fer phosphoprotein NCP94 PPP1R74 protein phosphatase 1 regulatory subunit 74 proto-oncogene c-Fer proto-oncogene tyrosine-protein kinase FER TYK3 Tyrosine kinase 3 tyrosine protein kinase FLK tyrosine-protein kinase Fer Tyrosine-protein kinase FLK; Feline encephalitis virus-related kinase FER; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p94-Fer; phosphoprotein NCP94; protein phosphatase 1, regulatory subunit 74; Proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; Tyrosine kinase 3; Tyrosine-protein kinase Fer
Gene Aliases: AV082135; C330004K01Rik; FER; Fert; Fert2; p94-Fer; PPP1R74; TYK3
UniProt ID: (Human) P16591, (Mouse) P70451
Entrez Gene ID: (Human) 2241, (Mouse) 14158
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.