Invitrogen
Manufacturer Code:PA538679
Catalog # PIPA538679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Moesin/Ezrin/Radixin around the phosphorylation site of Thr558 (Y-K-TP-L-R) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytovillin; cytovillin 2; epididymis luminal protein 70; epididymis secretory protein Li 105; ESP10; Ezrin; Membrane-organizing extension spike protein; Moesin; p81; Radixin; radixin isoform; villin 2; villin 2 (ezrin); Villin-2
Gene Aliases: AA516625; AW146364; C78546; CVIL; CVL; DFNB24; EZR; HEL-S-105; HEL70; MSN; p81; R75297; RDX; VIL2
UniProt ID: (Human) E1P5A8, (Mouse) P26040, (Rat) P31977, (Human) P26038, (Rat) O35763, (Mouse) P26041, (Human) P35241, (Mouse) P26043
Entrez Gene ID: (Human) 7430, (Mouse) 22350, (Rat) 54319, (Human) 4478, (Rat) 81521, (Mouse) 17698, (Human) 5962, (Rat) 315655, (Mouse) 19684
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.