Invitrogen
Manufacturer Code:PA537810
Catalog # PIPA537810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of tyrosine 324 (G-D-Y(p)-G-H) derived from Human Ephrin B1/B2/B3 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EFL-3; EFL6 EFNB3 EPH related receptor transmembrane ligand ELK L3 EPH related receptor tyrosine kinase ligand 8 Ephrin B3 EPLG8 LERK8 EFNB2 EPLG5 HTKL LERK5 Ephrin-B2 EPH-related receptor tyrosine kinase ligand 5 HTK ligand EFL6 EPLG8 LERK8 EPH-related receptor transmembrane ligand ELK-L3 Ephrin B3 eph-related receptor tyrosine kinase ligand 8; ELK ligand; ELK-L; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 5; EPH-related receptor tyrosine kinase ligand 8; Ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B2; Ephrin-B3; HTK ligand; HTK-L; LERK-2; LERK-5; LERK-8; ligand of eph-related kinase 5
Gene Aliases: CFND; CFNS; EFB1; EFL3; EFL6; EFNB1; EFNB2; EFNB3; Elk-L; EPLG2; EPLG5; EPLG8; Htk-L; HTKL; LERK2; LERK5; LERK8
UniProt ID: (Human) P98172, (Human) P52799, (Human) Q15768
Entrez Gene ID: (Human) 1947, (Human) 1948, (Human) 1949
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.