Invitrogen
Manufacturer Code:PA5103267
Catalog # PIPA5103267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Ephrin B1/B2/B3 around the phosphorylation site of Tyr324. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEK5 receptor ligand; CEK5 receptor ligand CEK5-L CFND CFNS EFB1 EFL3 EFL-3 EFL6 EFL-6 Efnb1 Efnb2 Efnb3 Elf2 ELF-2 ELF-3 ELK ligand ELK-L ELK-L3 EPH-related receptor transmembrane ligand ELK-L3 EPH-related receptor tyrosine kinase ligand 2 eph-related receptor tyrosine kinase ligand 5 EPH-related receptor tyrosine kinase ligand 8 ephrin B1 ephrin B2 ephrin B3 ephrin ligand B3 ephrin-B1 Ephrin-B1 C-terminal fragment Ephrin-B1 CTF Ephrin-B1 ICD Ephrin-B1 intracellular domain ephrin-B2 ephrin-B2 precursor ephrin-B3 Epl2 Epl5 Epl8 Eplg2 EPLG5 EPLG8 HTK ligand HTKL HTK-L Lerk2 LERK-2 Lerk5 LERK-5 LERK8 LERK-8 ligand of eph-related kinase 5 NLERK-1 NLERK-2 Stimulated by retinoic acid gene 1 protein Stra1; CEK5-L; EFL-3; ELF-2; ELK ligand; ELK-L; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 5; EPH-related receptor tyrosine kinase ligand 8; ephrin ligand B3; Ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B2; Ephrin-B3; HTK ligand; HTK-L; LERK-2; LERK-5; LERK-8; ligand of eph-related kinase 5; Stimulated by retinoic acid gene 1 protein
Gene Aliases: Cek5-L; CFND; CFNS; EFB1; EFL-3; EFL-6; EFL3; EFL6; EFNB1; EFNB2; EFNB3; ELF-2; ELF-3; Elf2; Elk-L; ELK-L3; Epl2; Epl5; Epl8; EPLG2; EPLG5; EPLG8; Htk-L; HTKL; LERK-2; LERK-5; LERK-8; LERK2; LERK5; LERK8; NLERK-1; NLERK-2; Stra1
UniProt ID: (Human) P98172, (Human) P52799, (Human) Q15768, (Mouse) P52795, (Mouse) P52800, (Mouse) O35393
Entrez Gene ID: (Human) 1947, (Human) 1948, (Human) 1949, (Rat) 25186, (Mouse) 13641, (Mouse) 13642, (Rat) 306636, (Mouse) 13643, (Rat) 360546
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.