Invitrogen
Manufacturer Code:PA537811
Catalog # PIPA537811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of tyrosine 588/596 (K-T-Y(p)-V-R)/(R-T-Y(p)-V-D) derived from Human EPHA2/3/4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARCC2 CTPA CTPP1 ECK ephrin type-A receptor 2 epithelial cell receptor protein tyrosine kinase soluble EPHA2 variant 1 tyrosine-protein kinase receptor ECK EK4 ETK ETK1 HEK HEK4 TYRO4 EPH-like kinase 4 TYRO4 protein tyrosine kinase eph-like tyrosine kinase 1 ephrin type-A receptor 3 human embryo kinase 1 tyrosine-protein kinase receptor ETK1 HEK8 SEK TYRO1 EK8 EPH-like kinase 8 TYRO1 protein tyrosine kinase ephrin type-A receptor 4 receptor protein-tyrosine kinase HEK8 tyrosine-protein kinase TYRO1 tyrosine-protein kinase receptor SEK; EK4; EK8; EPH-like kinase 4; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin type-A receptor 2; Ephrin type-A receptor 3; Ephrin type-A receptor 4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; HEK; Human embryo kinase; human embryo kinase 1; receptor protein-tyrosine kinase HEK8; soluble EPHA2 variant 1; testicular tissue protein Li 64; TYRO1 protein tyrosine kinase; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor MPK-5; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4
Gene Aliases: 2900005C20Rik; AI385584; ARCC2; AW492086; AW545284; Cek4; Cek8; CTPA; CTPP1; CTRCT6; ECK; EK4; End3; EPHA2; EPHA3; EPHA4; ETK; ETK1; HEK; HEK4; HEK8; Mek4; Myk2; rb; SEK; Sek-2; Sek1; Sek2; TYRO1; TYRO4
UniProt ID: (Human) P29317, (Human) P29320, (Human) P54764, (Mouse) Q03145, (Mouse) Q80VZ2
Entrez Gene ID: (Human) 1969, (Human) 2042, (Human) 2043, (Mouse) 13836, (Mouse) 13837, (Mouse) 13838
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.