Invitrogen
Manufacturer Code:MA515174
Catalog # PIMA515174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | B.742.5 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pThr202/Tyr204 of human p44 MAP kinase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; Diphospho-ERK; dp-ERK; dpERK; drosophila ERK; enhancer of seven in absentia 7; ERK; Erk MAP kinase; ERK-1; ERK-2; ERK/rolled; ERK2; ERT1; ERT2; extracellular signal-regulated kinase; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; Insulin-stimulated MAP2 kinase; MAP kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAP-kinase; MAPK; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase; mitogen activated protein kinase 1; mitogen-activated 3; mitogen-activated protein kinase; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 1 MAP kinase 1 MAPK 1 ERT1 Extracellular signal-regulated kinase 2 ERK-2 MAP kinase isoform p42 p42-MAPK Mitogen-activated protein kinase 2 MAP kinase 2 MAPK 2 MAPK1 ERK2 PRKM1 PRKM2 Mitogen-activated protein kinase 3 MAP kinase 3 MAPK 3 ERT2 Extracellular signal-regulated kinase 1 ERK-1 Insulin-stimulated MAP2 kinase MAP kinase isoform p44 p44-MAPK Microtubule-associated protein 2 kinase p44-ERK1 MAPK3 ERK1 PRKM3 p44 p42; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; Mitogen-activated protein kinase ERK-A; MNK1; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; phospho-ERK; pp42/MAP kinase; protein kinase; Protein rolled; protein tyrosine kinase ERK2; RE08694p; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; sevenmaker
Gene Aliases: 12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG18732; CT34260; CT39192; D-ERK; DERK; DERK-A; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmERK; DmERK-A; DmERKA; DmMAPK; dp-ERK; DpERK; dpERK1; dpMAPK; Dsor2; E(sina)7; EC2-1; EK2-1; ERK; ERK-1; ERK-2; ERK-A; Erk/Map kinase; ERK1; ERK1/2; ERK2; ERKA; ERT1; ERT2; Esrk1; EY2-2; fi06b09; GroupII; HS44KDAP; HUMKER1A; l(2)41Ac; l(2R)EMS45-39; MAP-k; MAPK; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; p-ERK; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; pERK; pMAPK; PRKM1; PRKM2; PRKM3; RL; rl/ERK; rl/MAPK; rll; SEM; SR2-1; Su(Raf)2B; wu:fi06b09; zERK1; zERK2
UniProt ID: (Human) P28482, (Human) P27361, (Mouse) P63085, (Rat) P63086, (Rat) P21708, (Mouse) Q63844, (Fruit fly) P40417
Entrez Gene ID: (Pig) 100153927, (Human) 5594, (Pig) 445013, (Human) 5595, (Mouse) 26413, (Rat) 116590, (Rat) 50689, (Mouse) 26417, (Zebrafish) 360144, (Zebrafish) 399480, (Fruit fly) 3354888
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.