Invitrogen
Manufacturer Code:PA5104552
Catalog # PIPA5104552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ERK1/2 around the phosphorylation site of Tyr204. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12559 9030612K14Rik AA407128 AU018647 BcDNA:RE08694 C78273 CG12559 CG12559 gene product from transcript CG12559-RC CG12559-PA CG12559-PC CG12559-PD CG12559-PE CG12559-PG CG12559-PH CG12559-PI CG18732 CT34260 CT39192 dERK D-ERK DERK-A Diphospho-ERK dm-dpERK DmelCG12559 Dmel_CG12559 DmErk DmERKA DmERK-A DmMAPK dpERk dp-ERK dpERK1 dpMAPK drosophila ERK Dsor2 E(sina)7 EC2-1 EK2-1 enhancer of seven in absentia 7 Erk Erk MAP kinase Erk/Map kinase ERK/rolled Erk1 ERK-1 erk1 erk2 ERK1b ERK2 ERK-2 ErkA ERK-A ERT1 Ert2 Esrk1 extracellular signal-regulated kinase extracellular signal-regulated kinase (ERK2) extracellular signal-regulated kinase 1 Extracellular signal-regulated kinase 2 extracellular signal-regulated kinase A extracellular signal-regulated kinase-1 extracellular signal-regulated kinase-2 extracellular signal-related kinase 1 Extracellular-regulated kinase A extracellular-signal-regulated kinase 2 Extracellular-signal-related kinase A EY2-2 fi06b09 GroupII HS44KDAP HUMKER1A I79_009500 I79_018350 Insulin-stimulated MAP2 kinase l(2)41Ac l(2R)EMS45-39 LOC100347345 LOC100348561 M phase MAP kinase MAP kinase MAP kinase 1 MAP kinase 2 MAP kinase 3 MAP kinase isoform p42 MAP kinase isoform p44 MapK MAP-k MAPK 1 MAPK 2 MAPK 3 MAPK1 mapk1.S mapk1a mapk1-a mapk1-b mapk2 MAPK3 MAP-kinase MBP kinase Microtubule-associated protein 2 kinase mitogen activated protein kinase mitogen activated protein kinase 1 mitogen activated protein kinase 3 mitogen activated protein kinase 3-like mitogen-activated 3 mitogen-activated protein kinase mitogen-activated protein kinase 1 mitogen-activated protein kinase 1 S homeolog mitogen-activated protein kinase 1a mitogen-activated protein kinase 1-like mitogen-activated protein kinase 2 Mitogen-activated protein kinase 3 Mitogen-activated protein kinase ERK-A MNK1 mpk1 Mtap2k Myelin basic protein kinase Myelin xP42 protein kinase p38 p40 p41 p41mapk p42 p42 MAP Kinase p42mapk p42-MAPK p44 p44 MAP kinase p44erk1 p44-ERK1 p44mapk p44-MAPK pERK phospho-ERK pMAPK pp42/MAP kinase Prkm1 prkm2 PRKM3 protein kinase Protein rolled protein tyrosine kinase ERK2 rl rl/ERK rl/MAPK rll rl-PA rl-PC rl-PD rl-PE rl-PG rl-PH rl-PI roll rolled sem sevenmaker SR2-1 Su(Raf)2B wu:fi06b09 XELAEV_18010534mg xp42 zERK1 zERK2; ERK-1; ERK-2; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-related kinase 1; extracellular-signal-regulated kinase 2; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 1; mitogen-activated 3; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; MNK1; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; pp42/MAP kinase; protein tyrosine kinase ERK2
Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Esrk1; HS44KDAP; HUMKER1A; Mapk; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3
UniProt ID: (Bovine) P46196, (Human) P28482, (Human) P27361, (Mouse) P63085, (Rat) P63086, (Rat) P21708, (Mouse) Q63844
Entrez Gene ID: (Bovine) 327672, (Human) 5594, (Bovine) 531391, (Human) 5595, (Mouse) 26413, (Rat) 116590, (Rat) 50689, (Mouse) 26417
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.