Invitrogen
Manufacturer Code:710213
Catalog # PI710213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 8HCLC |
Immunogen | Phosphopeptide corresponding to amino acids 11451153 of human epidermal growth factor receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; Epidermal growth factor receptor; Epidermal growth factor receptor mENA Oncogene ERBB Receptor tyrosine protein kinase ErbB1 Urogastrone wa2 Wa5 ERBB ERBB1 HER1 PIG61 mENA; erb-b2 receptor tyrosine kinase 1; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1
Gene Aliases: EGFR; ERBB; ERBB1; HER1; mENA; NISBD2; PIG61
UniProt ID: (Human) P00533
Entrez Gene ID: (Human) 1956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.