Invitrogen
Manufacturer Code:PA5105037
Catalog # PIPA5105037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DARPP-32 around the phosphorylation site of Thr75. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU040756 DARPP32 Darpp-32 dopamine- and cAMP-regulated neuronal phosphoprotein dopamine and cAMP-regulated neuronal phosphoprotein 32 dopamine- and cAMP-regulated phosphoprotein DARPP-32 FLJ20940 IPPD neuronal phosphoprotein DARPP-32 OTTHUMP00000164275 OTTHUMP00000164276 Ppp1r1b PPR1B protein pho protein phosphatase 1 regulatory inhibitor subunit 1B Protein phosphatase 1 regulatory subunit 1B protein phosphatase 1 regulatory (inhibitor) subunit 1B; DARPP-32; dopamine and cAMP-regulated neuronal phosphoprotein 32; Dopamine- and cAMP-regulated neuronal phosphoprotein; dopamine- and cAMP-regulated phosphoprotein DARPP-32; neuronal phosphoprotein DARPP-32; Protein phosphatase 1 regulatory subunit 1B
Gene Aliases: AU040756; DARPP-32; DARPP32; PPP1R1B
UniProt ID: (Human) Q9UD71, (Rat) Q6J4I0, (Mouse) Q60829
Entrez Gene ID: (Human) 84152, (Rat) 360616, (Mouse) 19049
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.