Invitrogen
Manufacturer Code:PA537541
Catalog # PIPA537541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of Serine 422(A-P-S(p)-P-G) derived from Human CtBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; BARS C-terminal-binding protein 1 brefeldin A-ribosylated substrate; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; CtBP1; CtBP3
Gene Aliases: 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; CTBP; CTBP1; CtBP1-L; CtBP1-S; Ctbp3; CtBP3/BARS; D4S115h; D5H4S115; D5H4S115E
UniProt ID: (Human) Q13363, (Rat) Q9Z2F5, (Mouse) O88712
Entrez Gene ID: (Human) 1487, (Rat) 29382, (Mouse) 13016
Molecular Function:
dehydrogenase
oxidoreductase
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.