Invitrogen
Manufacturer Code:PA537585
Catalog # PIPA537585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of serine 368 (R-A-S(p)-S-R) derived from Human Connexin 43. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 1 connexin; connexin 43; Connexin-43; Cx43; CX43 CXA1 CXN-43 Connexin 43 GJA1 Gap junction 43 kDa heart protein Gap junction alpha-1 protein; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction membrane channel protein alpha 1; gap junction protein, alpha 1, 43kDa
Gene Aliases: AU042049; AVSD3; AW546267; CMDR; Cnx43; connexin43; CX43; Cx43alpha1; Cxn-43; EKVP; Gja-1; GJA1; GJAL; HLHS1; HSS; Npm1; ODDD; PPKCA
UniProt ID: (Human) P17302, (Mouse) P23242, (Rat) P08050
Entrez Gene ID: (Human) 2697, (Mouse) 14609, (Rat) 24392
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.