Invitrogen
Manufacturer Code:PA517752
Catalog # PIPA517752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer3 of human cofilin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa phosphoprotein; 18 kDa phosphoprotein cofilin non-muscle isoform cofilin-1 p18; cofilin; cofilin 1 (non-muscle); Cofilin, non-muscle isoform; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; p18
Gene Aliases: AA959946; CFL; CFL1; Cof; COFILIN; Cofilin-1; HEL-S-15; n-cofilin
UniProt ID: (Human) P23528, (Bovine) Q5E9F7, (Pig) P10668, (Rat) P45592, (Mouse) P18760
Entrez Gene ID: (Human) 1072, (Bovine) 534553, (Pig) 445532, (Rat) 29271, (Mouse) 12631, (Chinese hamster) 100760435
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.