Invitrogen
Manufacturer Code:MA537003
Catalog # PIMA537003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | chk1S317-F10 |
Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: C85740 Cell cycle checkpoint kinase checkpoint kinase 1 checkpoint kinase 1 homolog checkpoint kinase-1 Checkpoint S. pombe homolog of 1 CHEK1 CHK1 CHK1 checkpoint homolog Chk1-S homolog of Chk1-S HRD1 Protein rad27 rad27 homolog serine/threonine-protein kinase Chk1 Synoviolin; Cell cycle checkpoint kinase; checkpoint kinase 1 homolog; Checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; rad27 homolog; Serine/threonine-protein kinase Chk1
Gene Aliases: C85740; CHEK1; CHK1; rad27
UniProt ID: (Human) O14757, (Mouse) O35280
Entrez Gene ID: (Human) 1111, (Mouse) 12649
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.