Invitrogen
Manufacturer Code:PA538433
Catalog # PIPA538433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human Calmodulin around the phosphorylation site of Thr79 and Ser81 (K-D-TP-D-SP-E-E) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM1 CALM2 PHKD PHKD3 caM calmodulin; calmodulin; Calmodulin 1 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1
Gene Aliases: AI256814; AI327027; AI461935; AL024000; CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMI; CaMII; CAMIII; CPVT4; DD132; LQT14; PHKD
UniProt ID: (Human) P0DP23, (Rat) P0DP29, (Mouse) P0DP26
Entrez Gene ID: (Human) 801, (Rat) 24242, (Mouse) 12313
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.