Invitrogen
Manufacturer Code:PA537833
Catalog # PIPA537833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide sequence around phosphorylation site of Threonine 287 (Q-E-T(p)-V-E) derived from Human CaMK2- beta/ gamma/ delta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta calcium/calmodulin-dependent protein kinase II delta calcium/calmodulin-dependent protein kinase type II delta chain Calcium/calmodulin-dependent protein kinase type II subunit delta Ca CAMKD CaM kinase II delta subunit CaM-kinase II delta chain CaMK-II delta subunit caM kinase II subunit delta caMK-II subunit delta calcium/calmodulin-dependent protein kinase (CaM kinase) II delta calcium/calmodulin-dependent protein kinase type II delta chain calcium/calmodulin-dependent protein kinase type II subunit delta CAMK CAMK-II CAMKG caMK-II subunit gamma calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma calcium/calmodulin-dependent protein kinase type II subunit gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; Calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit delta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II beta subunit; CaM kinase II delta subunit; CaM kinase II subunit beta; CaM kinase II subunit delta; CaM kinase II subunit gamma; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; CaMK II; CaMK-II delta subunit; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; proline rich calmodulin-dependent protein kinase
Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CAM2; CAMK; CaMK II; CAMK-II; CAMK1; CAMK2; CAMK2B; CAMK2D; CAMK2G; CAMKB; CAMKD; CAMKG; Camki; Ck2b; Kiaa4163; RATCAMKI
UniProt ID: (Human) Q13554, (Human) Q13557, (Human) Q13555, (Rat) P08413, (Mouse) P28652, (Rat) P15791, (Mouse) Q6PHZ2, (Mouse) Q923T9, (Rat) P11730
Entrez Gene ID: (Human) 816, (Human) 817, (Human) 818, (Rat) 24245, (Mouse) 12323, (Rat) 24246, (Mouse) 108058, (Mouse) 12325, (Rat) 171140
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.