Invitrogen
Manufacturer Code:PA538238
Catalog # PIPA538238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human CaMKII around the phosphorylation site of Thr305 (I-L-TP-T-M) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; Calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit alpha CaM kinase II subunit alpha CaMK-II subunit alpha CAMK2A CAMKA KIAA0968 CaMK2-a CaMK2a CaMK-2a Calcium/calmodulin-dependent protein kinase type II subunit delta CaM kinase II subunit delta CaMK-II subunit delta CAMK2D CAMKD CaMK2- delta CaMK2 delta CaMK-2 delta; Calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; CaM kinase II subunit alpha; CaM kinase II subunit beta; CaM kinase II subunit delta; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; CaMK II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; CaMKIINalpha; proline rich calmodulin-dependent protein kinase
Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CAM2; CaMK II; CAMK1; CAMK2; CAMK2A; CAMK2B; CAMK2D; CAMKA; CAMKB; CAMKD; Camki; CaMKII; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; R74975; RATCAMKI
UniProt ID: (Human) Q9UQM7, (Human) Q13554, (Human) Q13557, (Mouse) P11798, (Rat) P11275, (Rat) P08413, (Mouse) P28652, (Rat) P15791, (Mouse) Q6PHZ2
Entrez Gene ID: (Human) 815, (Human) 816, (Human) 817, (Mouse) 12322, (Rat) 25400, (Rat) 24245, (Mouse) 12323, (Rat) 24246, (Mouse) 108058
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.