Invitrogen
Manufacturer Code:PA538620
Catalog # PIPA538620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human IL-8R beta/CDw128 beta around the phosphorylation site of Ser347 (R-P-SP-F-V) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-X-C chemokine receptor type 2; CD182; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (C-X-C) receptor 2; chemokine (CXC) receptor 2; CXC-R2; CXCR-2; CXCR2 CDw128b CD182 antigen CMKAR2 CXCR-2 CD182 CXC-R2 IL-8R B Il-8 receptor b IL-8 receptor type 2 Il-8rb IL8R2 IL8RB IL-8 receptor beta; CXCR2 gene for IL8 receptor type B; GRO/MGSA receptor; High affinity interleukin-8 receptor B; IL-8 receptor alpha chain; IL-8 receptor type 2; IL-8R B; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B
Gene Aliases: CD128; CD182; CDw128; CDw128b; CMKAR2; CXCR2; Gpcr16; IL-8rb; IL-8Rh; IL8R2; IL8RA; IL8RB; mIL-8RH
UniProt ID: (Human) P25025, (Mouse) P35343
Entrez Gene ID: (Human) 3579, (Mouse) 12765
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.