Invitrogen
Manufacturer Code:MA515151
Catalog # PIMA515151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | F.540.2 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pTyr723 of human M-CSF receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: c-fms; CD115; CD115 antigen; CD115 antigen CSF-1-R FMS proto-oncogene McDonough feline sarcoma viral (v-fms) oncogene homolog OTTHUMP00000224093 macrophage colony stimulating factor I receptor macrophage colony-stimulating factor 1 receptor proto-oncogene c-Fms; CSF-1 receptor; CSF-1-R; FMS proto-oncogene; macrophage colony stimulating factor I receptor; Macrophage colony-stimulating factor 1 receptor; McDonough feline sarcoma viral (v-fms) oncogene homolog; Proto-oncogene c-Fms; proto-oncogene fms
Gene Aliases: AI323359; C-FMS; CD115; CSF-1R; CSF1R; Csfmr; CSFR; Fim-2; FIM2; FMS; HDLS; M-CSF-R; M-CSFR
UniProt ID: (Human) P07333, (Mouse) P09581
Entrez Gene ID: (Human) 1436, (Mouse) 12978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.