Invitrogen
Manufacturer Code:MA515145
Catalog # PIMA515145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | S.48.4 |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer345 of human Chk1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle checkpoint kinase; checkpoint kinase 1 homolog; Checkpoint kinase-1; Checkpoint S. pombe homolog of 1 serine/threonine-protein kinase Chk1; Checkpoint, S. pombe, homolog of, 1; CHK1 checkpoint homolog; Chk1-S; rad27 homolog; Serine/threonine-protein kinase Chk1
Gene Aliases: C85740; CHEK1; CHK1; rad27
UniProt ID: (Human) O14757, (Rat) Q91ZN7, (Mouse) O35280
Entrez Gene ID: (Human) 1111, (Rat) 140583, (Mouse) 12649
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.